Assay Details
| Gene Symbol: NUDT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334912.1 | NP_001320544.1 | 13 - 14 | 1536 | 88 | |
| NM_106618.5 | NP_565218.1 | 12 - 13 | 1708 | 88 | ||
| GenBank mRNA | AY120774.1 | - | 12 - 13 | 1424 | 88 | |
| BT000142.1 | - | 12 - 13 | 1394 | 88 | ||
More Information
Set Membership: |
|

NUDT3
Genomic Map