Assay Details
Gene Symbol: GL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198514.1 | NP_001185443.1 | 2 - 3 | 300 | 59 | |
NM_001334927.1 | NP_001320268.1 | 1 - 2 | 171 | 59 | ||
NM_001334928.1 | NP_001320269.1 | 1 - 2 | 171 | 59 | ||
NM_106633.2 | NP_565223.1 | 1 - 2 | 226 | 59 | ||
GenBank mRNA | AF360294.1 | - | 1 - 2 | 187 | 59 | |
AJ457046.1 | - | 1 - 2 | 76 | 59 | ||
BT001956.1 | - | 1 - 2 | 76 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
