Assay Details
Gene Symbol: GL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198514.1 | NP_001185443.1 | 4 - 5 | 693 | 69 | |
NM_001334926.1 | NP_001320267.1 | 3 - 4 | 795 | 69 | ||
NM_001334927.1 | NP_001320268.1 | 3 - 4 | 564 | 69 | ||
NM_001334928.1 | NP_001320269.1 | 3 - 4 | 564 | 69 | ||
NM_001334929.1 | NP_001320270.1 | 2 - 3 | 457 | 69 | ||
NM_106633.2 | NP_565223.1 | 3 - 4 | 619 | 69 | ||
GenBank mRNA | AF360294.1 | - | 3 - 4 | 580 | 69 | |
AJ457046.1 | - | 3 - 4 | 469 | 69 | ||
BT001956.1 | - | 3 - 4 | 469 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
