Assay Details
| Gene Symbol: ROPGEF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106635.4 | NP_178104.1 | 2 - 3 | 748 | 97 | |
| GenBank mRNA | BT033042.1 | - | 2 - 3 | 543 | 97 | |
More Information
Set Membership: |
|

ROPGEF12
Genomic Map