Assay Details
Gene Symbol: IMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106704.4 | NP_178171.1 | 1 - 2 | 281 | 90 | |
GenBank mRNA | Y10216.1 | - | 254 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
