Assay Details
| Gene Symbol: RTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106713.4 | NP_178180.1 | 3 - 4 | 516 | 117 | |
| GenBank mRNA | AK117343.1 | - | 3 - 4 | 460 | 117 | |
| BT005219.1 | - | 3 - 4 | 429 | 117 | ||
More Information
Set Membership: |
|

RTL1
Genomic Map