Assay Details
| Gene Symbol: CCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106730.4 | NP_178197.1 | 3 - 4 | 982 | 66 | |
| GenBank mRNA | AK227576.1 | - | 3 - 4 | 883 | 66 | |
| BT005826.1 | - | 3 - 4 | 810 | 66 | ||
More Information
Set Membership: |
|

CCR2
Genomic Map