Assay Details
| Gene Symbol: UBC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337764.1 | NP_001319500.1 | 2 - 3 | 233 | 79 | |
| NM_111704.2 | NP_566332.1 | 1 - 2 | 111 | 79 | ||
| GenBank mRNA | BT024821.1 | - | 1 - 2 | 66 | 79 | |
| DQ027026.1 | - | 1 - 2 | 66 | 79 | ||
More Information
Set Membership: |
|

UBC12
Genomic Map