Assay Details
| Gene Symbol: RAPTOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111719.3 | NP_566335.1 | 20 - 21 | 4020 | 89 | |
| GenBank mRNA | AY059838.1 | - | 20 - 21 | 3920 | 89 | |
| AY769948.1 | - | 20 - 21 | 3920 | 89 | ||
| BT008813.1 | - | 20 - 21 | 3531 | 89 | ||
More Information
Set Membership: |
|

RAPTOR1
Genomic Map