Assay Details
| Gene Symbol: RAPTOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111719.3 | NP_566335.1 | 3 - 4 | 989 | 99 | |
| GenBank mRNA | AY059838.1 | - | 3 - 4 | 889 | 99 | |
| AY769948.1 | - | 3 - 4 | 889 | 99 | ||
| BT008813.1 | - | 3 - 4 | 500 | 99 | ||
More Information
Set Membership: |
|

RAPTOR1
Genomic Map