Assay Details
| Gene Symbol: RVE8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337828.1 | NP_001327347.1 | 6 - 7 | 1310 | 140 | |
| NM_001337832.1 | NP_001327348.1 | 7 - 8 | 967 | 140 | ||
| NM_001337833.1 | NP_001327346.1 | 6 - 7 | 1199 | 140 | ||
| NM_111794.4 | NP_187571.2 | 7 - 8 | 1023 | 140 | ||
| GenBank mRNA | AY091106.1 | - | 7 - 8 | 847 | 140 | |
| AY122969.1 | - | 7 - 8 | 803 | 140 | ||
More Information
Set Membership: |
|

RVE8
Genomic Map