Assay Details
Gene Symbol: SYP71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111809.5 | NP_566354.1 | 7 - 8 | 971 | 112 | |
GenBank mRNA | AF355757.1 | - | 7 - 8 | 749 | 112 | |
AY090358.1 | - | 7 - 8 | 817 | 112 | ||
BT024483.1 | - | 7 - 8 | 712 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
