Assay Details
| Gene Symbol: ADK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111817.4 | NP_187593.1 | 1 - 2 | 429 | 76 | |
| NM_202540.1 | NP_974269.1 | 1 - 2 | 115 | 76 | ||
| GenBank mRNA | AF180894.1 | - | 1 - 2 | 136 | 76 | |
| AF375451.1 | - | 1 - 2 | 156 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_202540.1 transcripts.
Set Membership: |
|

ADK1
Genomic Map