Assay Details
Gene Symbol: OMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111840.3 | NP_187616.1 | 2 - 3 | 897 | 71 | |
GenBank mRNA | AF096281.1 | - | 2 - 3 | 590 | 71 | |
AY065037.1 | - | 2 - 3 | 609 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
