Assay Details
Gene Symbol: OMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111840.3 | NP_187616.1 | 7 - 8 | 1808 | 129 | |
GenBank mRNA | AF096281.1 | - | 7 - 8 | 1501 | 129 | |
AY065037.1 | - | 7 - 8 | 1520 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
