Assay Details
| Gene Symbol: AT3G10060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111841.5 | NP_187617.1 | 7 - 8 | 758 | 73 | |
| GenBank mRNA | AY094442.1 | - | 7 - 8 | 608 | 73 | |
| AY122895.1 | - | 7 - 8 | 541 | 73 | ||
More Information
Set Membership: |
|

AT3G10060
Genomic Map