Assay Details
Gene Symbol: CAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337885.1 | NP_001325725.1 | 2 - 3 | 831 | 73 | |
NM_111896.2 | NP_187671.1 | 3 - 4 | 1212 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
