Assay Details
Gene Symbol: FER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111942.4 | NP_187716.1 | 4 - 5 | 700 | 100 | |
GenBank mRNA | AJ312192.1 | - | 4 - 5 | 559 | 100 | |
AK175807.1 | - | 4 - 5 | 580 | 100 | ||
AK175899.1 | - | 4 - 5 | 580 | 100 | ||
AK175901.1 | - | 4 - 5 | 580 | 100 | ||
AK175910.1 | - | 4 - 5 | 580 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
