Assay Details
| Gene Symbol: AT3G11100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111947.4 | NP_566386.1 | 1 - 2 | 1314 | 103 | |
| GenBank mRNA | AY064137.1 | - | 1 - 2 | 785 | 103 | |
More Information
Set Membership: |
|

AT3G11100
Genomic Map