Assay Details
Gene Symbol: AT3G11130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111950.3 | NP_187724.2 | 29 - 30 | 5360 | 76 | |
GenBank mRNA | AK229443.1 | - | 29 - 30 | 5167 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
