Assay Details
| Gene Symbol: CASP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111988.3 | NP_187762.1 | 1 - 2 | 360 | 61 | |
| GenBank mRNA | BT010843.1 | - | 1 - 2 | 342 | 61 | |
| BT012612.1 | - | 1 - 2 | 276 | 61 | ||
More Information
Set Membership: |
|

CASP2
Genomic Map