Assay Details
| Gene Symbol: HMG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113794.4 | NP_189515.1 | 10 - 11 | 1218 | 130 | |
| GenBank mRNA | BT002507.1 | - | 10 - 11 | 1212 | 130 | |
More Information
Set Membership: |
|

HMG
Genomic Map