Assay Details
| Gene Symbol: KIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113839.4 | NP_566843.1 | 9 - 10 | 1498 | 112 | |
| NM_202645.3 | NP_974374.1 | 9 - 10 | 1350 | 112 | ||
| NM_202646.1 | NP_974375.1 | 9 - 10 | 1271 | 112 | ||
| GenBank mRNA | AY070468.1 | - | 9 - 10 | 1301 | 112 | |
| AY149927.1 | - | 8 - 9 | 1198 | 112 | ||
| DQ778956.1 | - | 8 - 9 | 1198 | 112 | ||
| X99279.1 | - | 1284 | 112 | |||
| Gene Symbol: AT2G15960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230337.1 | - | 447 | 112 | ||
More Information
Set Membership: |
|

KIN11
Genomic Map