Assay Details
| Gene Symbol: AT3G30200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113919.4 | NP_189639.4 | 1 - 2 | 74 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G30200| Gene Symbol: AT3G30200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113919.4 | NP_189639.4 | 1 - 2 | 74 | 98 | |
Set Membership: |
|