Assay Details
| Gene Symbol: ERD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339059.1 | NP_001319672.1 | 2 - 3 | 909 | 74 | |
| NM_113981.6 | NP_189701.3 | 1 - 2 | 997 | 74 | ||
More Information
Set Membership: |
|

ERD5
Genomic Map