Assay Details
Gene Symbol: HRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114479.5 | NP_190196.1 | 2 - 3 | 448 | 82 | |
GenBank mRNA | AF020715.1 | - | 2 - 3 | 384 | 82 | |
AY056233.1 | - | 2 - 3 | 389 | 82 | ||
AY117252.1 | - | 2 - 3 | 379 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
