Assay Details
| Gene Symbol: AT3G46480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114515.4 | NP_190232.3 | 2 - 3 | 223 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G46480| Gene Symbol: AT3G46480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114515.4 | NP_190232.3 | 2 - 3 | 223 | 97 | |
Set Membership: |
|