Assay Details
Gene Symbol: TRFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035744.2 | NP_001030821.1 | 5 - 6 | 1547 | 69 | |
NM_001035745.2 | NP_001030822.2 | 5 - 6 | 1214 | 69 | ||
NM_001339266.1 | NP_001326037.1 | 5 - 6 | 1324 | 69 | ||
NM_001339267.1 | NP_001326038.1 | 3 - 4 | 845 | 69 | ||
NM_114526.3 | NP_190243.2 | 5 - 6 | 1229 | 69 | ||
GenBank mRNA | AJ630475.1 | - | 4 - 5 | 853 | 69 | |
AK229705.1 | - | 5 - 6 | 1136 | 69 | ||
AY395985.1 | - | 5 - 6 | 1185 | 69 | ||
AY519541.1 | - | 4 - 5 | 853 | 69 | ||
AY568647.1 | - | 4 - 5 | 853 | 69 | ||
BT033142.1 | - | 4 - 5 | 856 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
