Assay Details
Gene Symbol: TRFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035744.2 | NP_001030821.1 | 7 - 8 | 1818 | 78 | |
NM_001035745.2 | NP_001030822.2 | 7 - 8 | 1485 | 78 | ||
NM_001339267.1 | NP_001326038.1 | 5 - 6 | 1116 | 78 | ||
NM_114526.3 | NP_190243.2 | 7 - 8 | 1500 | 78 | ||
GenBank mRNA | AJ630475.1 | - | 6 - 7 | 1124 | 78 | |
AK229705.1 | - | 7 - 8 | 1407 | 78 | ||
AY395985.1 | - | 7 - 8 | 1456 | 78 | ||
AY519541.1 | - | 6 - 7 | 1124 | 78 | ||
AY568647.1 | - | 6 - 7 | 1124 | 78 | ||
BT033142.1 | - | 6 - 7 | 1127 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
