Assay Details
| Gene Symbol: PHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203097.2 | NP_001190026.1 | 8 - 9 | 1417 | 117 | |
| NM_114607.3 | NP_190323.1 | 8 - 9 | 1417 | 117 | ||
More Information
Set Membership: |
|

PHS1
Genomic Map