Assay Details
| Gene Symbol: KING1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114711.2 | NP_190422.1 | 3 - 4 | 716 | 106 | |
| GenBank mRNA | AJ132317.1 | - | 3 - 4 | 645 | 106 | |
| AY042839.1 | - | 3 - 4 | 627 | 106 | ||
| AY081464.1 | - | 3 - 4 | 535 | 106 | ||
More Information
Set Membership: |
|

KING1
Genomic Map