Assay Details
Gene Symbol: CDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114734.4 | NP_566911.1 | 3 - 4 | 460 | 85 | |
GenBank mRNA | AK226373.1 | - | 3 - 4 | 358 | 85 | |
AY085153.1 | - | 3 - 4 | 85 | |||
BT024706.1 | - | 1 - 2 | 191 | 85 | ||
M59198.1 | - | 3 - 4 | 85 | |||
S45387.1 | - | 3 - 4 | 85 | |||
X57839.1 | - | 349 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
