Assay Details
Gene Symbol: CDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114734.4 | NP_566911.1 | 7 - 8 | 1007 | 123 | |
GenBank mRNA | AK226373.1 | - | 7 - 8 | 905 | 123 | |
AY085153.1 | - | 7 - 8 | 907 | 123 | ||
BT024706.1 | - | 5 - 6 | 738 | 123 | ||
M59198.1 | - | 7 - 8 | 905 | 123 | ||
S45387.1 | - | 7 - 8 | 905 | 123 | ||
X57839.1 | - | 896 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
