Assay Details
Gene Symbol: CDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114734.4 | NP_566911.1 | 8 - 9 | 1060 | 73 | |
GenBank mRNA | AK226373.1 | - | 8 - 9 | 958 | 73 | |
AY085153.1 | - | 8 - 9 | 960 | 73 | ||
BT024706.1 | - | 6 - 7 | 791 | 73 | ||
M59198.1 | - | 8 - 9 | 958 | 73 | ||
S45387.1 | - | 8 - 9 | 958 | 73 | ||
X57839.1 | - | 949 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
