Assay Details
Gene Symbol: ABIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035755.2 | NP_001030832.1 | 3 - 4 | 497 | 88 | |
NM_114788.4 | NP_190498.1 | 3 - 4 | 750 | 88 | ||
GenBank mRNA | AY817013.1 | - | 2 - 3 | 162 | 88 | |
BT026081.1 | - | 2 - 3 | 162 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
