Assay Details
Gene Symbol: ABIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035755.2 | NP_001030832.1 | 7 - 8 | 883 | 67 | |
NM_114788.4 | NP_190498.1 | 7 - 8 | 1136 | 67 | ||
GenBank mRNA | AY817013.1 | - | 6 - 7 | 548 | 67 | |
BT026081.1 | - | 6 - 7 | 548 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
