Assay Details
| Gene Symbol: CID10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035756.2 | NP_001030833.1 | 3 - 4 | 852 | 128 | |
| NM_001339409.1 | NP_001325461.1 | 3 - 4 | 782 | 128 | ||
| NM_114799.4 | NP_190508.1 | 3 - 4 | 826 | 128 | ||
| GenBank mRNA | AK317338.1 | - | 3 - 4 | 810 | 128 | |
More Information
Set Membership: |
|

CID10
Genomic Map