Assay Details
| Gene Symbol: DIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114822.5 | NP_190531.2 | 1 - 2 | 190 | 82 | |
| GenBank mRNA | BT000898.1 | - | 1 - 2 | 114 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DIN11| Gene Symbol: DIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114822.5 | NP_190531.2 | 1 - 2 | 190 | 82 | |
| GenBank mRNA | BT000898.1 | - | 1 - 2 | 114 | 82 | |
Set Membership: |
|