Assay Details
Gene Symbol: AT3G49740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114834.3 | NP_190543.1 | 1 | 2257 | 89 |
Gene Symbol: AT3G49730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339438.1 | NP_001319717.1 | 1 | 201 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001339438.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
