Assay Details
| Gene Symbol: PEN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114871.3 | NP_566929.1 | 10 - 11 | 1718 | 130 | |
| GenBank mRNA | BT000931.1 | - | 9 - 10 | 1256 | 130 | |
More Information
Set Membership: |
|

PEN3
Genomic Map