Assay Details
| Gene Symbol: CEJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114886.3 | NP_190595.1 | 1 | 663 | 74 | |
| GenBank mRNA | AY045579.1 | - | 1 | 588 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEJ1| Gene Symbol: CEJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114886.3 | NP_190595.1 | 1 | 663 | 74 | |
| GenBank mRNA | AY045579.1 | - | 1 | 588 | 74 | |
Set Membership: |
|