Assay Details
Gene Symbol: SNRK2.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203118.1 | NP_001190047.1 | 8 - 9 | 1072 | 111 | |
NM_114910.3 | NP_190619.1 | 8 - 9 | 1068 | 111 | ||
GenBank mRNA | BT025246.1 | - | 8 - 9 | 798 | 111 | |
L05561.1 | - | 8 - 9 | 942 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
