Assay Details
| Gene Symbol: DWF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339479.1 | NP_001327730.1 | 2 - 3 | 780 | 119 | |
| NM_114926.4 | NP_190635.1 | 2 - 3 | 780 | 119 | ||
| GenBank mRNA | AF412114.1 | - | 2 - 3 | 571 | 119 | |
| AY090266.1 | - | 2 - 3 | 543 | 119 | ||
More Information
Set Membership: |
|

DWF4
Genomic Map