Assay Details
| Gene Symbol: AT3G50880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114948.4 | NP_190657.1 | 1 - 2 | 1031 | 117 | |
| GenBank mRNA | AK119087.1 | - | 1 - 2 | 855 | 117 | |
More Information
Set Membership: |
|

AT3G50880
Genomic Map