Assay Details
| Gene Symbol: TRX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114963.5 | NP_190672.1 | 2 - 3 | 373 | 65 | |
| GenBank mRNA | AY088687.1 | - | 2 - 3 | 231 | 65 | |
| Z14084.1 | - | 248 | 65 | |||
More Information
Set Membership: |
|

TRX1
Genomic Map