Assay Details
Gene Symbol: F3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203121.1 | NP_001190050.1 | 2 - 3 | 421 | 106 | |
NM_114983.3 | NP_190692.1 | 1 - 2 | 573 | 106 | ||
GenBank mRNA | AF428335.1 | - | 1 - 2 | 389 | 106 | |
AY058886.1 | - | 1 - 2 | 395 | 106 | ||
AY116957.1 | - | 1 - 2 | 356 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
