Assay Details
| Gene Symbol: AT3G51390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339514.1 | NP_001326942.1 | 10 - 11 | 1018 | 76 | |
| NM_114998.5 | NP_566950.1 | 10 - 11 | 1032 | 76 | ||
| GenBank mRNA | BT010141.1 | - | 10 - 11 | 859 | 76 | |
More Information
Set Membership: |
|

AT3G51390
Genomic Map