Assay Details
| Gene Symbol: AT3G51390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339514.1 | NP_001326942.1 | 3 - 4 | 445 | 79 | |
| NM_114998.5 | NP_566950.1 | 3 - 4 | 444 | 79 | ||
| GenBank mRNA | BT010141.1 | - | 3 - 4 | 271 | 79 | |
More Information
Set Membership: |
|

AT3G51390
Genomic Map