Assay Details
| Gene Symbol: DET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117086.4 | NP_192756.2 | 9 - 10 | 1622 | 61 | |
| GenBank mRNA | BT015348.1 | - | 9 - 10 | 1525 | 61 | |
| BT020454.1 | - | 9 - 10 | 1490 | 61 | ||
More Information
Set Membership: |
|

DET1
Genomic Map