Assay Details
| Gene Symbol: AT4G10260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117094.3 | NP_192764.1 | 4 - 5 | 861 | 65 | |
| GenBank mRNA | DQ446815.1 | - | 4 - 5 | 842 | 65 | |
More Information
Set Membership: |
|

AT4G10260
Genomic Map